Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Beclin 2 Polyclonal Antibody, Invitrogen™

Catalog No. PIPA577774
Change view
Click to view available options
Quantity:
100 μg
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
PIPA577774 100 μg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. PIPA577774 Supplier Thermo Scientific Supplier No. PA577774
Add to Cart
Add to Cart

Rabbit Polyclonal Antibody

A 1:1,000 dilution of PA5-77774 was sufficient for detection of Beclin2 on rat liver lysates using Goat anti-rabbit IgG:HRP as the secondary antibody.∣Predicted molecular weight at approximately 48.1kDa. Observed bands in the 50-55kDa range, varied by lysates.

Involved in 2 distinct lysosomal degradation pathways: acts as a regulator of autophagy and as a regulator of G-protein coupled receptors turnover. Regulates degradation in lysosomes of a variety of G-protein coupled receptors via its interaction with GPRASP1/GASP1. [UniProt]
TRUSTED_SUSTAINABILITY

Specifications

Antigen Beclin 2
Applications Immunocytochemistry, Western Blot
Classification Polyclonal
Concentration 1 mg/mL
Conjugate Unconjugated
Formulation PBS with 50% glycerol and 0.09% sodium azide; pH 7.4
Gene Becn2
Gene Accession No. A8MW95
Gene Alias Bcl-2; BCS2; beclin 1, autophagy related, pseudogene 1; beclin 2; beclin-1 autophagy-related pseudogene 1; Beclin-1-like protein 1; Beclin-2; BECN1L1; BECN1P1; BECN2; Gm104; PFM 2; PFM2; PRDM 5; PRDM5; PRDM5 protein; RGD1563293; similar to Beclin 1 (Coiled-coil myosin-like BCL2-interacting protein)
Gene Symbols Becn2
Host Species Rabbit
Immunogen Synthetic peptide from the mid-protein of human Beclin 2 (C-DQQERQHYRDYS)
Purification Method Antigen affinity chromatography
Quantity 100 μg
Regulatory Status RUO
Primary or Secondary Primary
Gene ID (Entrez) 289269, 441925
Target Species Human, Rat
Content And Storage -20°C
Product Type Antibody
Form Liquid
Isotype IgG
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.