Learn More
Invitrogen™ Beclin 2 Polyclonal Antibody

Description
Suggested positive control: A431 and HEK kidney cell lysates and MCC827 lung cell lysates, A-431 whole cell lysate1, 293 whole cell lysate.
Specifications
Specifications
| Antigen | Beclin 2 |
| Applications | Western Blot |
| Classification | Polyclonal |
| Concentration | 0.9 mg/mL |
| Conjugate | Unconjugated |
| Formulation | PBS with 30% glycerol and 0.1% sodium azide |
| Gene | Becn2 |
| Gene Accession No. | A8MW95 |
| Gene Alias | Bcl-2; BCS2; beclin 1, autophagy related, pseudogene 1; beclin 2; beclin-1 autophagy-related pseudogene 1; Beclin-1-like protein 1; Beclin-2; BECN1L1; BECN1P1; BECN2; Gm104; PFM 2; PFM2; PRDM 5; PRDM5; PRDM5 protein; RGD1563293; similar to Beclin 1 (Coiled-coil myosin-like BCL2-interacting protein) |
| Gene Symbols | Becn2 |
| Show More |
By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.