Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Beclomethasone Dipropionate, Micronized, USP, 97-103%, Spectrum™ Chemical
SDP

Catalog No. 18600860 Shop All Spectrum Chemical Manufacturing Corporation Products
Click to view available options
Quantity:
250 mg
1 g
5 g
100 g
1 kg
Packaging:
Amber Glass Bottle

B1412, 5534-09-8, C28H37ClO7

Spectrum™ Chemical Beclomethasone Dipropionate, Micronized, USP is potent glucocorticoid steroid used to prevent and control symptoms caused by asthma. All Spectrum Chemical USP grade products are manufactured, packaged and stored under current Good Manufacturing Practices (cGMP) per 21CFR part 211 in FDA registered and inspected facilities.

Spectrum™ Chemical Cholecalciferol, Crystal, USP is also known as toxiferol and is a form of vitamin D. It is used in vitamin supplements. All Spectrum Chemical USP products are manufactured, packaged and stored under current Good Manufacturing Practices (cGMP) per 21CFR part 211 in FDA registered and inspected facilities.

Chemical Identifiers

CAS 5534-09-8
Molecular Formula C28H37ClO7
Molecular Weight (g/mol) 521.05
InChI Key KUVIULQEHSCUHY-XYWKZLDCSA-N
IUPAC Name 2-[(1R,2S,3aS,3bS,9aS,9bR,10S,11aS)-9b-chloro-10-hydroxy-2,9a,11a-trimethyl-7-oxo-1-(propanoyloxy)-1H,2H,3H,3aH,3bH,4H,5H,7H,9aH,9bH,10H,11H,11aH-cyclopenta[a]phenanthren-1-yl]-2-oxoethyl propanoate
SMILES CCC(=O)OCC(=O)[C@@]1(OC(=O)CC)[C@@H](C)C[C@H]2[C@@H]3CCC4=CC(=O)C=C[C@]4(C)[C@@]3(Cl)[C@@H](O)C[C@]12C

Specifications

CAS Max % 100%
Melting Point 201°C to 208°C
Ignition Residue 0.1%
Loss on Drying 0.5%
Packaging Amber Glass Bottle
Quantity 250 mg
Specific Rotation +88° to +94°
Percent Purity 97 to 103%
Grade USP
Physical Form Powder or Waxy Solid
Chemical Name or Material Beclomethasone Dipropionate
Show More Show Less
WARNING: Reproductive Harm - www.P65Warnings.ca.gov
Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Your feedback has been submitted: Thank you for helping us improve our website.