Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Beclomethasone Dipropionate, MP Biomedicals™

Catalog No. ICN15481483 Shop All MP Biomedicals, Inc Products
Click to view available options
Quantity:
250 mg
1 g
5 g

Catalog No. ICN15481483

Supplier: MP Biomedicals, Inc 0215481483

Item Discontinued This item has been discontinued by the supplier. Please to view product availability in your area.

Antiallergic, antiasthmatic, anti-inflammatory
TRUSTED_SUSTAINABILITY

Specifications

CAS 9-8-5534
Color White
Molecular Formula C28H37ClO7
Quantity 250 mg
Synonym beclomethasone dipropionate, beclometasone dipropionate, beconase, beclovent, vancenase, beclazone, becloforte, beclomet, beclorhinol, propaderm
InChI Key KUVIULQEHSCUHY-XYWKZLDCSA-N
SMILES CCC(=O)OCC(=O)C1(C(CC2C1(CC(C3(C2CCC4=CC(=O)C=CC43C)Cl)O)C)C)OC(=O)CC
IUPAC Name [2-[(8R,10S,11S,13S,14S,16S,17R)-9-chloro-11-hydroxy-10,13,16-trimethyl-3-oxo-17-propanoyloxy-6,7,8,11,12,14,15,16-octahydrocyclopenta[a]phenanthren-17-yl]-2-oxoethyl] propanoate
Molecular Weight (g/mol) 521.05
PubChem CID 134129500
Formula Weight 521
Physical Form Solid
Chemical Name or Material Beclomethasone Dipropionate
Show More Show Less
storageNote1 Protect from light.
Recommended Storage Store at +4°C
Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Your feedback has been submitted: Thank you for helping us improve our website.