Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More
Learn More
BECN1 Monoclonal Antibody (OTI3H8), TrueMAB™, OriGene

Mouse Monoclonal Antibody
Supplier: OriGene TA502517S


Description
BECN1 Monoclonal antibody specifically detects BECN1 in Human samples. It is validated for Flow Cytometry, Immunocytochemistry, Immunohistochemistry (Paraffin), Western BlotSpecifications
BECN1 | |
Monoclonal | |
0.55 mg/mL | |
PBS with 1% BSA, 50% glycerol and 0.02% sodium azide | |
Q9H227 | |
Mouse | |
Affinity chromatography | |
RUO | |
57733 | |
-20° C, Avoid Freeze/Thaw Cycles | |
IgG2a |
Flow Cytometry, Immunocytochemistry, Immunohistochemistry (Paraffin), Western Blot | |
OTI3H8 | |
Unconjugated | |
GBA3 | |
4921513J16Rik; 5430417M23Rik; APG6; Atg6; ATG6 autophagy related 6 homolog; Bcl-2-interacting protein beclin; beclin 1; beclin 1 (coiled-coil, moesin-like BCL2-interacting protein); beclin 1 (coiled-coil, myosin-like BCL2 interacting protein); beclin 1 (coiled-coil, myosin-like BCL2-interacting protein); beclin 1, autophagy related; beclin1; beclin-1; Beclin-1; beclin-1; Beclin-1-C 35 kDa; Beclin-1-C 37 kDa; becn1; Coiled-coil myosin-like BCL2-interacting protein; coiled-coil, myosin-like BCL2-interacting protein; GT197; hypothetical protein MGC73097; myosin-like BCL2 interacting protein; Protein GT197; testis secretory sperm-binding protein Li 215e; VPS30; zgc:73097 | |
Human recombinant protein fragment corresponding to amino acids 1-150 and 370-469 of human GBA3 produced in E.coli. | |
30 μL | |
Primary | |
Human | |
Liquid |
Product Content Correction
The Fisher Scientific Encompass Program offers items which are not part of our distribution portfolio. These products typically do not have pictures or detailed descriptions. However, we are committed to improving your shopping experience. Please use the form below to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction