Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

BECN1 Mouse anti-Human, Clone: 15CT26, Abnova™

Catalog No. 89933639 Shop All Abnova Corporation Products
Change view
Click to view available options
Quantity:
100 μL
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
89-933-639 100 μL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. 89-933-639 Supplier Abnova Corporation Supplier No. MAB12283
Add to Cart
Add to Cart

Mouse monoclonal antibody raised against full length recombinant human BECN1.

Specifications

Antigen BECN1
Applications Immunofluorescence, Immunohistochemistry (PFA fixed), Western Blot
Classification Monoclonal
Clone 15CT26
Conjugate Unconjugated
Description Mouse monoclonal antibody raised against full length recombinant human BECN1.
Dilution Immunohistochemistry (1:50-1:200) Immunofluorescence (1:100) Western Blot (1:50-1:2000) The optimal working dilution should be determined by the end user.
Formulation In ascites (0.09% sodium azide)
Gene Alias ATG6/VPS30/beclin1
Gene Symbols BECN1
Host Species Mouse
Immunogen Recombinant His fusion protein corresponding to full length human BECN1.
Quantity 100 μL
Regulatory Status RUO
Primary or Secondary Primary
Gene ID (Entrez) 8678
Target Species Human
Content And Storage Store at 4°C. For long term storage store at -20°C.
Aliquot to avoid repeated freezing and thawing.
Form Liquid
Isotype IgG3 λ
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.