Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Invitrogen™ TFPI (Befovacimab Biosimilar) Recombinant Human Monoclonal Antibody
GREENER_CHOICE

Catalog No. PIMA542266
Change view
Click to view available options
Quantity:
100 μg
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
PIMA542266 100 μg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
This item is not returnable. View return policy
Catalog No. PIMA542266 Supplier Invitrogen™ Supplier No. MA542266
Add to Cart
Add to Cart
This item is not returnable. View return policy

Human Recombinant Monoclonal Antibody

Endotoxin level <0.01EU/μg by LAL method Use a manual defrost freezer and avoid repeated freeze thaw cycles. Store at 2 to 8 °C for one week. Store at -20 to -80 °C for twelve months from the date of receipt. This product is a Humanized mouse antibody.

Tissue factor pathway inhibitor (TFPI), also known as lipoprotein-associated coagulation inhibitor, is a protease inhibitor that regulates the tissue factor (TF)-dependent pathway of blood coagulation. TFPI is glycosylated and predominantly found in the vascular endothelium and plasma in both free forms and complexed with plasma lipoproteins. The coagulation process initiates with the formation of a factor VIIa-TF complex, which proteolytically activates additional proteases (factors IX and X) and ultimately leads to the formation of a fibrin clot. TFPI inhibits the activated factor X and VIIa-TF proteases in an autoregulatory loop.
TRUSTED_SUSTAINABILITY

Specifications

Antigen TFPI (Befovacimab Biosimilar)
Applications ELISA, Flow Cytometry
Classification Recombinant Monoclonal
Concentration 1 mg/mL
Conjugate Unconjugated
Formulation PBS with no preservative; pH 7.4
Gene TFPI
Gene Accession No. P10646
Gene Alias A630013F22Rik; anti-convertin; AW552122; EPI; Extrinsic pathway inhibitor; LACI; Lipoprotein-associated coagulation inhibitor; OTTHUMP00000163472; OTTHUMP00000205431; TFI; TFPI; TFPI1; Tissue factor pathway inhibitor; tissue factor pathway inhibitor (lipoprotein-associated coagulation inhibitor)
Gene Symbols TFPI
Host Species Human
Immunogen Human TFPI.
Purification Method Protein A
Quantity 100 μg
Regulatory Status RUO
Primary or Secondary Primary
Gene ID (Entrez) 7035
Target Species Human
Content And Storage -20°C, Avoid Freeze/Thaw Cycles
Product Type Antibody
Form Liquid
Isotype IgG2 λ
Target Molecule Chemical
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.