Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More
Learn More
BEGAIN Monoclonal Antibody (OTI6E7), TrueMAB™, OriGene

Mouse Monoclonal Antibody
Supplier: OriGene CF810836


Description
BEGAIN Monoclonal antibody specifically detects BEGAIN in Human samples. It is validated for Western BlotSpecifications
BEGAIN | |
Monoclonal | |
See Label | |
PBS with 8% trehalose and no preservative; pH 7.3 | |
P09919 | |
Mouse | |
Protein A/G | |
RUO | |
1440 | |
-20° C, Avoid Freeze/Thaw Cycles | |
IgG1 |
Western Blot | |
OTI6E7 | |
Unconjugated | |
Csf3 | |
BEGAIN; BM948371; brain enriched guanylate kinase associated; brain-enriched guanylate kinase-associated; brain-enriched guanylate kinase-associated homolog; brain-enriched guanylate kinase-associated protein; brain-enriched guanylate kinase-associated; brain-enriched guanylate kinase-associated protein; Gm897; KIAA1446 | |
Full length human recombinant protein of human CSF3 produced in HEK293T. | |
100 μg | |
Primary | |
Human | |
Lyophilized |
Product Content Correction
The Fisher Scientific Encompass Program offers items which are not part of our distribution portfolio. These products typically do not have pictures or detailed descriptions. However, we are committed to improving your shopping experience. Please use the form below to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction