Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Invitrogen™ BEGAIN Polyclonal Antibody
GREENER_CHOICE

Catalog No. PIPA531568
Change view
Click to view available options
Quantity:
100 μL
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
PIPA531568 100 μL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. PIPA531568 Supplier Invitrogen™ Supplier No. PA531568
Add to Cart
Add to Cart

Rabbit Polyclonal Antibody

Recommended positive controls: Raji. Predicted reactivity: Mouse (95%), Rat (95%), Sheep (85%), Bovine (86%). Store product as a concentrated solution. Centrifuge briefly prior to opening the vial.

May sustain the structure of the postsynaptic density (PSD). Target protein binds the guanylate kinase domain of PSD-95/SAP90, may play a role in the post-synaptic membrane.
TRUSTED_SUSTAINABILITY

Specifications

Antigen BEGAIN
Applications Immunohistochemistry (Paraffin), Western Blot
Classification Polyclonal
Concentration 1 mg/mL
Conjugate Unconjugated
Formulation 0.1M tris glycine with 20% glycerol and 0.01% thimerosal; pH 7
Gene BEGAIN
Gene Accession No. Q9BUH8
Gene Alias BEGAIN; BM948371; brain enriched guanylate kinase associated; brain-enriched guanylate kinase-associated; brain-enriched guanylate kinase-associated homolog; brain-enriched guanylate kinase-associated protein; brain-enriched guanylate kinase-associated; brain-enriched guanylate kinase-associated protein; Gm897; KIAA1446
Gene Symbols BEGAIN
Host Species Rabbit
Immunogen Recombinant protein encompassing a sequence within the center region of human BEGAIN. The exact sequence is proprietary.
Purification Method Antigen affinity chromatography
Quantity 100 μL
Regulatory Status RUO
Primary or Secondary Primary
Gene ID (Entrez) 57596
Target Species Human
Content And Storage Store at 4°C short term. For long term storage, store at -20°C, avoiding freeze/thaw cycles.
Product Type Antibody
Form Liquid
Isotype IgG
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.