Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More
Learn More
Invitrogen™ BEGAIN Polyclonal Antibody

Rabbit Polyclonal Antibody
Supplier: Invitrogen™ PA551769
Description
Immunogen sequence: PYPAETFRFP ASPGPQQALM PPNLWSLRAK PGTARLPGED MRGQWRPLSV EDIGAYSYPV SAAGRASPCS FSERYYGGAG GSPGKKADGR ASPLYASYKA DSFSEGDDLS QGHLAEPCFL RAGGDLSLSP GRSADPLPGY APSE Highest antigen sequence identity to the following orthologs: Mouse - 78%, Rat - 81%.
May sustain the structure of the postsynaptic density (PSD). Target protein binds the guanylate kinase domain of PSD-95/SAP90, may play a role in the post-synaptic membrane.
Specifications
| BEGAIN | |
| Polyclonal | |
| Unconjugated | |
| BEGAIN | |
| BEGAIN; BM948371; brain enriched guanylate kinase associated; brain-enriched guanylate kinase-associated; brain-enriched guanylate kinase-associated homolog; brain-enriched guanylate kinase-associated protein; brain-enriched guanylate kinase-associated; brain-enriched guanylate kinase-associated protein; Gm897; KIAA1446 | |
| Rabbit | |
| Antigen Affinity Chromatography | |
| RUO | |
| 57596 | |
| Store at 4°C short term. For long term storage, store at -20°C, avoiding freeze/thaw cycles. | |
| Liquid |
| Immunohistochemistry (Paraffin), Western Blot | |
| 0.1 mg/mL | |
| PBS with 40% glycerol and 0.02% sodium azide; pH 7.2 | |
| Q9BUH8 | |
| BEGAIN | |
| Recombinant protein corresponding to Human BEGAIN. Recombinant protein control fragment (Product #RP-89071). | |
| 100 μL | |
| Primary | |
| Human | |
| Antibody | |
| IgG |
Safety and Handling
WARNING: Cancer - www.P65Warnings.ca.gov
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction