Learn More
Bel Art™ Battery-Powered Engraving Tool and Stand
Catalog No. p-4122870 Shop All SP Bel-Art & SP Wilmad-LabGlass Products
Change view
Click to view available options
Material:
Plastic
Polypropylene
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Material
08-647-870 Plastic
08-647-871 Polypropylene
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
This item is not returnable. View return policy
Catalog No. 08-647-870 Supplier Bel Art™ Supplier No. F441500005
Only null left
Add to Cart
Add to Cart
This item is not returnable. View return policy
Includes
Extra engraving tip
Requires
AA batteries (not included)
Permanently mark important tools, samples and more in seconds
  • Hold the Engraving Tool like a pen, press the power button, apply light pressure, and write to permanently mark most materials including glass, plastic, wood, metal, bone, and stone
  • Portable, cordless battery power: operates on two AA batteries (not included)
  • Use indoors or out in the field
  • Marks will not fade if exposed to chemicals or sunlight
  • Engraving Tool Stand (Mfr. No. F441500015)
  • Keeps tool handy when not in use
  • Molded from polyethylene
  • Well to hold extra engraving tip
Order Info
7 in. (17.8cm) long
Specifications
Product Type Battery-powered Engraving Tool
For Use With (Equipment) Engrave test tubes, bottles, racks, keys, equipment, tools, electronics and more
Length (English) 7 in.
Length (Metric) 18 cm
Material Plastic
Includes Extra engraving tip