Learn More
Bel-Art™ Polyethylene Carboy with Spigot
Catalog No. 22290047 Shop All SP Bel-Art & SP Wilmad-LabGlass Products
Click to view available options
Capacity (Metric):
4 L
20 L
For dispensing of cleaning solutions, laboratory chemicals, disinfectants, and intermediate phase mixtures and slurries for various manufacturing processes.
  • Lighter than glass and easy to handle.
  • Heavy knurling on cap provides excellent grip for opening and closing.
  • Resistant to corrosive acids and alkalis.
  • Spigot requires only half turn to open or close.
  • Smooth sides accept adhesive labels and can be marked with most indelible markers.
  • Use to dispense cleaning solutions, laboratory reagents, disinfectants, and intermediate phase mixtures and slurries for various manufacturing processes.
Specifications
Autoclavable Not Autoclavable
Capacity (English) 5.3 gal.
Capacity (Metric) 20 L
Closure Material HDPE
Closure Type Threaded
Color White
Disposable Reusable
For Use With (Application) For dispense cleaning solutions, laboratory chemicals, disinfectants, and intermediate phase mixtures and slurries for various manufacturing processes
Includes Spigot
Material HDPE
Mouth Style Narrow
Neck Style Threaded
Product Type Carboy with Spigot
Shape Cylindrical
Sterility Non-sterile
Show More Show Less
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Your feedback has been submitted: Thank you for helping us improve our website.