Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Bel-Art™ Cleanware™ Self-Adhesive Waste Bags

Catalog No. 0181511F Shop All SP Bel-Art & SP Wilmad-LabGlass Products
Change view
Click to view available options
Dimensions (L x W):
10 x 8 in.
16 x 12 in.
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Dimensions (L x W)
01-815-11F 10 x 8 in.
01-815-11G 16 x 12 in.
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. 01-815-11F Supplier Bel-Art™ Supplier No. F131741008
Only null left
Add to Cart
Add to Cart

Disposable waste bags free bench or hood space when hung from walls. Bel Art™ Cleanware™ Self-Adhesive Waste Bags adhere to cabinets and other surfaces.

  • Peel backing off the adhesive strip across the top and hang close to your work area
  • Back panel is longer than the front for easy opening
  • High Molecular Weight High Density (HMHD) polyethylene; steam autoclavable at 121°C (250°F)
  • Maximum weight of contents should not exceed 1.36kg (3lb)

Specifications

Autoclavable Autoclavable
Closure Type Zip Tie
Color White
Disposable Single-use
For Use With (Application) Cleanware
For Use With (Equipment) General Trash
Includes Self-adhesive strip
Material HDPE
Sterility Non-sterile
Thickness (English) 1 mil
Dimensions (L x W) 10 x 8 in.
Length (English) 10 in.
Width (English) 8 in.
Product Type Laboratory Waste Bag
Capacity (English) 0.5 gal.
Capacity (Metric) 2 L
Length (Metric) 25.4 cm
Width (Metric) 20.32 cm
Thickness (Metric) 0.025 mm
Show More Show Less
WARNING: Cancer - www.P65Warnings.ca.gov
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.