Learn More
Bel-Art™ Economy Frigimat™ Dry-Ice Maker
GSA_VA
Catalog No. 22261841 Shop All SP Bel-Art & SP Wilmad Products
Change view
Click to view available options
Material:
Aluminum, Brass, Epoxy-coated Steel
Catalog No. Material
22-261841 Aluminum, Brass, Epoxy-coated Steel
1 options
Catalog No. 22-261841 Supplier Bel-Art™ Supplier No. H388760000
Get Up to $500 in Products with Purchase
View all promotions
Add to Cart
Add to Cart
Each 50 lb. cylinder of liquid CO2 will produce 8 or 9 blocks of dry ice measuring 3.62 x 4 in. (8.5 x 11 cm) long
  • Low-density polyethylene (LDPE) ice chamber is held in place by metal clamping mechanism
  • Presure gauge with relief valve ensures safe operation by minimizing pressure build up
  • Unit constructed of epoxy coated steel, aluminum and brass
  • Includes a 1.8m (6ft) nickel plated copper connecting tube with standard CGA 320 (0.830 in.-14) thread couplings for easy attachment to a LCO2 cylinder
  • Has four non-skid rubber feet to prevent sliding
This product(s) resides on a Fisher Scientific GSA or VA contract. If you are viewing this page as a nonregistered user, the price(s) displayed is List Price. To view your GSA or VA contract pricing, log in using your account number, or become a registered user by contacting one of our Customer Service teams. You can also view your contract price by searching for this item(s) on GSA Advantage. To place an order, contact Fisher Scientific Customer Service.
Specifications
Product Type Dry Ice Maker
Type Dry Ice Maker
Material Aluminum, Brass, Epoxy-coated Steel
Yield 16 oz. in 1 to 2 min.
Holds 8.5 x 11cm L 8 to 9 blocks of dry ice
For Use With (Application) Ideal for use in remote locations or low dry ice demand
Includes 1.8m Nickel plated Copper connecting tube with standard CGA 320 (0.83 in. to 14) thread couplings, non-skid Rubber feet, pressure gauge with relief valve
For Use With (Equipment) 50 lb. LCO2 cylinder