Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Bel Art™ PiRack™ Pipetter Holder System

Catalog No. p-3205768
Click to view available options
Product Type:
Extenders
Pipet Tray
Single Clamp Holder
Triple Clamp Holder
This item is not returnable. View return policy
This item is not returnable. View return policy

Mix and match components to suit your needs

  • Polypropylene system is strong enough to support the weight of even the heaviest pipettor
  • Holds a single pipette with one clamp or add a triple clamp or extenders to increase the number of holders at a work location
  • Utility tray mounts between two clamps, ideal for storing pipetting supplies or other small items
  • Perfectly sized for No Wire™ Half Racks that hold tubes, vials, mini tubes and cuvettes (series 18748)
  • Attach to any shelf or benchtop edge up to 2.54 cm (1 in.) thick using the large locking thumbscrew clamp
  • Supports many standard brands of pipettors
  • All components are chemical resistant and autoclavable at 121°C (250°F)

Specifications

Product Type Extenders
Array 1 x 1
Autoclavable Autoclavable
Color Blue
Holds Pipets
Material Polypropylene
Positions 1
Shape Rectangular
Shelves None
Stackable No
Sterility Non-sterile
For Use With (Equipment) Standard brands of pipettors
Show More Show Less
Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Your feedback has been submitted: Thank you for helping us improve our website.