Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Bel-Art™ Red Wash Bottles

Catalog No. 011890107 Shop All SP Bel-Art & SP Wilmad-LabGlass Products
Click to view available options
Capacity (Metric):
250 mL
500 mL
Mouth:
Narrow
Wide

The bright red color helps identify hazardous contents at a glance.

  • Low-density polyethylene with polypropylene closure

Specifications

Capacity 8 oz., 250 mL
Capacity (Metric) 250 mL
Capacity (English) 8 oz.
Color Red
Material LDPE
Sterility Non-sterile
Closure Included Yes
Autoclavable Not Autoclavable
Disposable Reusable
Graduated No
Mouth Narrow
Shape Cylindrical
Product Type Economy Wash Bottle
Chemical Label Blank
Closure Material Polypropylene
Closure Size 28 mm
Show More Show Less
Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Your feedback has been submitted: Thank you for helping us improve our website.