Learn More
Bel-Art™ SP Scienceware™ Replacement Photoelectric Cells for Klett Colorimeters
Shop All SP Bel-Art & SP Wilmad-LabGlass ProductsDescription
- Uses existing colorimeter hardware
- Includes two cells (must be replaced as pair)
Specifications
Specifications
| Product Type | Photoelectric Cell |
| For Use With (Equipment) | Klett™ 37012-0000, T37012-0001, T37013-0000 and T37013-0001 colorimeters |
| Includes | Two cells |