Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More
Learn More
Bel Art™ Secador™ Auto Desiccator Cabinet, Vertical Model
Shop All SP Bel-Art & SP Wilmad-LabGlass Products
Click to view available options
Color:
Amber
Blue with Clear Door
Clear
Clear with Blue Ends
Cabinet Style:
Vertical
Vertical (Tinted)
Voltage:
120 V
230 V
Volume (English):
1.16 cu. ft.
1.55 cu. ft.
1.87 cu. ft.
Description
A UV light blocking, dry environment with automatic electronic dehumidification.
- Attractive design and sturdy construction of the Secador line of desiccator cabinets assures easy and reliable storage of moisture and light sensitive products.
- Patented electronic dehumidification system reduces humidity and circulates dry air automatically, freeing users from monitoring and changing desiccants.
- Durastar™ co-polyester construction blocks 99% of UV light and resists staining, crazing and chemical attack.
- Amber color models reduce visible light penetration by over 50% for storing light sensitive materials like reagents, analytical standards and investigational compounds.
- Large doors with easy-to-view dial hygrometer maximize access to interior space and have a patented seal design, tab latches and security loops for a padlock or tamper-evident seals.
- Perforated shelves facilitate circulation of dry air; Space for desiccant under bottom shelf.
- 4.0 horizontal has a wider footprint, lower height and pull down oven style door.
- U.S. Patents: 6,772,534; 6,834,920; 7,114,266; 7,318,630 and D474,626.
- 4.0 units can be converted from horizontal to vertical configuration or vice versa by turning the unit and ordering the appropriate number of shelves (see Ordering Info).
- Secador 2.0: 1.16 cu. ft. internal volume, 16.3 L x 13.4 W x 12.4 in. H
- Secador 3.0: 1.55 cu. ft. internal volume, 16.3 L x 13.4 W x 16.4 in. H
- Secador 4.0: 1.87 cu. ft. internal volume, 16.3 L x 13.4 W x 20.4 in. H
Order Info
Replacement shelves are available.
Specifications
Specifications
Autoclavable | Not Autoclavable |
Color | Amber |
Cabinet Style | Vertical (Tinted) |
Material | Copolyester |
Product Type | Desiccator |
Door Style | Side-swinging |
For Use With (Application) | Easy and reliable storage of moisture and light sensitive products, can be used with Scienceware™ desiccant cartridges, canisters or similar desiccant |
Includes | Carrying handles, 2 latches and loops for a padlock or tamper-evident seals, door-mounted hygrometer |
Model | 4.0 |
No. of Shelves | 3 |
Show More |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction