Learn More
Invitrogen™ BCMA (Belantamab Biosimilar) Recombinant Human Monoclonal Antibody

Description
Endotoxin level <0.01EU/μg by LAL method Use a manual defrost freezer and avoid repeated freeze thaw cycles. Store at 2 to 8 °C for one week. Store at -20 to -80 °C for twelve months from the date of receipt. This product is a Humanized mouse antibody.
Specifications
Specifications
| Antigen | BCMA (Belantamab Biosimilar) |
| Applications | ELISA |
| Classification | Recombinant Monoclonal |
| Concentration | 1.31 mg/mL |
| Conjugate | Unconjugated |
| Formulation | PBS with no preservative; pH 7.4 |
| Gene | Tnfrsf17 |
| Gene Accession No. | Q02223 |
| Gene Alias | B cell maturation antigen; B-cell maturation factor; B-cell maturation protein; BCM; BCMA; CD269; TNF receptor superfamily member 17; Tnfrsf13; Tnfrsf13a; Tnfrsf17; Tumor necrosis factor receptor superfamily member 17; tumor necrosis factor receptor superfamily, member 17 |
| Gene Symbols | Tnfrsf17 |
| Show More |
By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.