Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Invitrogen™ Belimumab Recombinant Human Monoclonal Antibody
GREENER_CHOICE

Catalog No. PIMA541860
Change view
Click to view available options
Quantity:
100 μg
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
PIMA541860 100 μg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
This item is not returnable. View return policy
Catalog No. PIMA541860 Supplier Invitrogen™ Supplier No. MA541860
Add to Cart
Add to Cart
This item is not returnable. View return policy

Human Recombinant Monoclonal Antibody

Endotoxin level < 0.01EU/μg by LAL method Use a manual defrost freezer and avoid repeated freeze thaw cycles. Store at 2 to 8 °C for one week. Store at -20 to -80 °C for twelve months from the date of receipt.

Belimumab is an intravenous immunosuppressant for the adjunctive treatment of systemic lupus erythematosus (SLE). More specifically, it is a fully human recombinant IgG1lambda monoclonal antibody produced from a recombinant NS0 cell line stably transfected with the belimumab heavy chain and light chain genes. It is the first biological treatment approved for the indication of SLE. Concomitant use with live or inactivated vaccines must be avoided. Belimumab was FDA approved on March 9, 2011. Belimumab consists of 2 heavy chains, and 2 light chains of the lambda subclass. Each heavy chain contains 452 amino acid residues and each light chain contains 214 amino acid residues. There are 3 post-translational modifications: a conserved N-linked glycosylation on the CH2 domain at Asn 303 of the heavy chain, the conversion of the N-terminal glutamine residue of the heavy chain into pyroglutamate, and loss of C-terminal lysine residue of the heavy chain.
TRUSTED_SUSTAINABILITY

Specifications

Antigen Belimumab
Applications ELISA, SDS-Page
Classification Recombinant Monoclonal
Concentration 1.73 mg/mL
Conjugate Unconjugated
Formulation PBS with no preservative; pH 7.4
Gene Alias hBlySmAb-1.1; LymphoStat-B
Host Species Human
Immunogen Human TNFSF13B/CD257.
Purification Method Protein A
Quantity 100 μg
Regulatory Status RUO
Primary or Secondary Primary
Target Species Human
Content And Storage -20°C, Avoid Freeze/Thaw Cycles
Product Type Antibody
Form Liquid
Isotype IgG1 λ
Target Molecule Chemical
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.