Description
- Cover the top buffer tray to protect from buffer spillages and salt deposits
- Simplify daily and weekly preventive maintenance of the top buffer tray of ÄKTA systems
- Manufactured of absorbent, impermeable material that is designed to protect lab surfaces against hazardous spills
- Smooth, absorbent Whatman paper quickly absorbs spills
- Laminated polyethylene layer prevents flow-through to the working surface
- Fits the top buffer tray on ÄKTA go, ÄKTA avant, ÄKTA pure and ÄKTA start protein purification systems.
- Convenient and easy to use
Specifications
Specifications
| Product Type | Absorbent Liner |
| Material | Paper, Laminated Polyethylene |
| For Use With (Application) | ÄKTA Start |
| Quantity | 50/Pk. |