Learn More
Cytiva Benchkote Sheet for ÄKTA Systems
Catalog No. 09928150 Shop All Cytiva Products
Change view
Click to view available options
For Use With (Application):
ÄKTA Avant
ÄKTA Pure
ÄKTA Start
Quantity:
10/Pk.
25/Pk.
50/Pk.
9 product options available for selection
Product selection table with 9 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. For Use With (Application) Quantity
09-928-150 ÄKTA Avant 25/Pk.
09-928-149 ÄKTA Avant 10/Pk.
09-928-151 ÄKTA Avant 50/Pk.
09-928-143 ÄKTA Pure 10/Pk.
09-928-144 ÄKTA Pure 25/Pk.
09-928-145 ÄKTA Pure 50/Pk.
09-928-146 ÄKTA Start 10/Pk.
09-928-147 ÄKTA Start 25/Pk.
09-928-148 ÄKTA Start 50/Pk.
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 9 options available.
9 options
Catalog No. 09-928-150 Supplier Cytiva Supplier No. 230010073
Add to Cart
Add to Cart
Protects the top buffer trays from buffer spillages and salt deposits
  • Cover the top buffer tray to protect from buffer spillages and salt deposits
  • Simplify daily and weekly preventive maintenance of the top buffer tray of ÄKTA systems
  • Manufactured of absorbent, impermeable material that is designed to protect lab surfaces against hazardous spills
  • Smooth, absorbent Whatman paper quickly absorbs spills
  • Laminated polyethylene layer prevents flow-through to the working surface
  • Fits the top buffer tray on ÄKTA go, ÄKTA avant, ÄKTA pure and ÄKTA start protein purification systems.
  • Convenient and easy to use
TRUSTED_SUSTAINABILITY
Specifications
Product Type Absorbent Liner
Material Paper, Laminated Polyethylene
For Use With (Application) ÄKTA Avant
Quantity 25/Pk.