Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More
Learn More
Sartorius Benchtop Meters: pHBasic+ Series
Click to view available options
Type:
pH Meter with Kit
pH Meter with Kit and Electrode
Description
- Fast and accurate results, easy operation, and capacity to have direct data recall and transfer
- Full functionality with USB data transmission
- Recall and output of 5,000 data set memory
- Easy-to-use menus and intuitive operation sets
- Sensor efficiency & calibration integrity
- Clear status icons and indicators
- Large LCD display
- Automatic temperature compensation
- Automatic buffer recognition
- Programmable data collection
- 5-point calibration gives a broader range in slope accuracy
- Features Continuous Monitoring Control CMC, GLP TIME/DATE stamps

Specifications
Specifications
Description | Meter, Power Supply, Cover, Arm, 3 Buffer, Electrode: PY-P10-2S (5pt Cal, ElectCheck, USB,CMC) |
Includes | Power Supply, Manual, 4 AA batteries, Electrode Arm and Base, Electrode PY-P10-2S |
Outputs | USB |
For Use With (Application) | Sample and Buffer Preparation - Regulated R&D and QC environments |
Calibration Points | 5 |
Display Type | LCD |
Memory | Data Memory 500 and output |
Warranty | 2 Years |
Model | pHBasic+ |
Type | pH Meter with Kit and Electrode |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction