Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Invitrogen™ BEND2 Polyclonal Antibody
GREENER_CHOICE

Catalog No. PIPA552669
Change view
Click to view available options
Quantity:
100 μL
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
PIPA552669 100 μL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. PIPA552669 Supplier Invitrogen™ Supplier No. PA552669
Add to Cart
Add to Cart

Rabbit Polyclonal Antibody

Immunogen sequence: GLKQFFHLSL PSSWDDRRTP PCPVAHGDQI VSQINHPVHL RRYSYNSEEV DFPKRGRFYT PEVQSSISPP AERQETHAWA SPAVTSLESA ACHELQEADL SESLSYPRIV SSSSLQQYVA QGGSFPCFGM PWNFISGG Highest antigen sequence identity to the following orthologs: Mouse - 21%, Rat - 21%.

The BEND2 gene, located on chromosome Xp22.2, encodes a protein that is part of the BEN domain-containing family, which is implicated in chromatin organization and transcriptional regulation. BEND2 is critical in spermatogenesis, particularly in the meiotic phase of germ cell development. Knockout studies in male mice have shown that loss of BEND2 disrupts the transition from zygonema to pachynema, interferes with chromosomal synapsis, and impairs the repair of DNA double-strand breaks, ultimately causing infertility. Additionally, BEND2's interaction with chromatin-associated proteins suggests its role in the formation of transcription-repressor complexes, essential for proper gene expression regulation during meiosis.
TRUSTED_SUSTAINABILITY

Specifications

Antigen BEND2
Applications Immunohistochemistry (Paraffin), Western Blot
Classification Polyclonal
Concentration 0.2 mg/mL
Conjugate Unconjugated
Formulation PBS with 40% glycerol and 0.02% sodium azide; pH 7.2
Gene BEND2
Gene Accession No. Q8NDZ0
Gene Alias BEN domain containing 2; BEN domain-containing protein 2; BEND2; CXorf20
Gene Symbols BEND2
Host Species Rabbit
Immunogen Recombinant protein corresponding to Human BEND2. Recombinant protein control fragment (Product #RP-92246).
Purification Method Antigen affinity chromatography
Quantity 100 μL
Regulatory Status RUO
Primary or Secondary Primary
Gene ID (Entrez) 139105
Target Species Human
Content And Storage Store at 4°C short term. For long term storage, store at -20°C, avoiding freeze/thaw cycles.
Product Type Antibody
Form Liquid
Isotype IgG
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.