Learn More
Invitrogen™ BEND2 Polyclonal Antibody
Rabbit Polyclonal Antibody
Supplier: Invitrogen™ PA553022
Description
Immunogen sequence: DQDDASASAC LTPDFALLPL NILVKVDTNT ENSVNTMNRS TLLDSDSGQD SSSSSVCIPP KYGYLGDPKR NVRVLKIHLL AVQNMAKPKQ AACYLVRILF SKEILISSSV DIHLKDSQSL DPNKMAALRE YLATTFPTCD LHEHG Highest antigen sequence identity to the following orthologs: Mouse - 32%, Rat - 21%.

Specifications
BEND2 | |
Polyclonal | |
Unconjugated | |
BEND2 | |
BEN domain containing 2; BEN domain-containing protein 2; BEND2; CXorf20 | |
Rabbit | |
Antigen affinity chromatography | |
RUO | |
139105 | |
Store at 4°C short term. For long term storage, store at -20°C, avoiding freeze/thaw cycles. | |
Liquid |
Immunohistochemistry (Paraffin), Western Blot | |
0.10 mg/mL | |
PBS with 40% glycerol and 0.02% sodium azide; pH 7.2 | |
Q8NDZ0 | |
BEND2 | |
Recombinant protein corresponding to Human BEND2. Recombinant protein control fragment (Product #RP-92245). | |
100 μL | |
Primary | |
Human | |
Antibody | |
IgG |
Your input is important to us. Please complete this form to provide feedback related to the content on this product.