Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More
Learn More
Invitrogen™ BEND4 Polyclonal Antibody
Rabbit Polyclonal Antibody
Supplier: Invitrogen™ PA5101533
Description
Antibody detects endogenous levels of total BEND4.
BEND4, also named as CCDC4, has 5 isoforms with MW 55-58 kDa, 45-48 kDa and 36 kDa.
Specifications
BEND4 | |
Polyclonal | |
Unconjugated | |
BEND4 | |
BEN domain containing 4; BEN domain-containing protein 4; BEN domain-containing protein 4-like; bend4; CCDC4; coiled-coil domain containing 4; coiled-coil domain-containing protein 4; D330027G24Rik; EG666938 | |
Rabbit | |
Affinity Chromatography | |
RUO | |
389206, 681008 | |
-20°C | |
Liquid |
Western Blot, Immunocytochemistry | |
1 mg/mL | |
PBS with 50% glycerol and 0.02% sodium azide; pH 7.4 | |
Q6ZU67 | |
BEND4 | |
A synthesized peptide derived from human BEND4(Accession Q6ZU67), corresponding to amino acid residues D378-L428. | |
100 μL | |
Primary | |
Human, Rat | |
Antibody | |
IgG |
Safety and Handling
WARNING: Cancer - www.P65Warnings.ca.gov
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction