Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Invitrogen™ BEND4 Polyclonal Antibody
GREENER_CHOICE

Catalog No. PIPA557846
Change view
Click to view available options
Quantity:
100 μL
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
PIPA557846 100 μL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. PIPA557846 Supplier Invitrogen™ Supplier No. PA557846
Add to Cart
Add to Cart

Rabbit Polyclonal Antibody

Immunogen sequence: QYMQRKQQTS AFLRVFTDSL QNYLLSGSFP TPNPSSASEY GHLADVDPLS TSPVHTLGGW TSPATSESHG HPSSSTL Highest antigen sequence identity to the following orthologs: Mouse - 97%, Rat - 95%.

BEND4, also named as CCDC4, has 5 isoforms with MW 55-58 kDa, 45-48 kDa and 36 kDa.
TRUSTED_SUSTAINABILITY

Specifications

Antigen BEND4
Applications Immunohistochemistry (Paraffin), Immunocytochemistry
Classification Polyclonal
Concentration 0.3 mg/mL
Conjugate Unconjugated
Formulation PBS with 40% glycerol and 0.02% sodium azide; pH 7.2
Gene BEND4
Gene Accession No. Q6ZU67
Gene Alias BEN domain containing 4; BEN domain-containing protein 4; BEN domain-containing protein 4-like; bend4; CCDC4; coiled-coil domain containing 4; coiled-coil domain-containing protein 4; D330027G24Rik; EG666938
Gene Symbols BEND4
Host Species Rabbit
Immunogen Recombinant protein corresponding to Human BEND4. Recombinant protein control fragment (Product #RP-96753).
Purification Method Antigen Affinity Chromatography
Quantity 100 μL
Regulatory Status RUO
Primary or Secondary Primary
Gene ID (Entrez) 389206
Target Species Human
Content And Storage Store at 4°C short term. For long term storage, store at -20°C, avoiding freeze/thaw cycles.
Product Type Antibody
Form Liquid
Isotype IgG
Show More Show Less
WARNING: Cancer - www.P65Warnings.ca.gov
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.