Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Benedict Quantitative Solution, Certified, 1.95 to 2.05mg/mL equivalence, LabChem™

Catalog No. p-5388366
Click to view available options
Quantity:
500 mL
1 L

Specifications

Chemical Name or Material Benedict Quantitative Solution
Color Blue
CAS Max % 58.21, 20.04, 12.52, 7.41, 1.8
CAS 63126-89-6, 333-20-0, 497-19-8, 7758-99-8
Linear Formula CuSO4·5H2O
Concentration 1.95 to 2.05mg/mL equivalence
Solubility Information Soluble in water
Formula Weight 249.68
Grade Certified
Identification Passes Test
Packaging Poly Bottle
Decomposition Information Hydrogen cyanide; Carbon monoxide; Carbon dioxide; Sulfur compounds; Copper
Quantity 500 mL
Physical Form Liquid
Show More Show Less
Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Your feedback has been submitted: Thank you for helping us improve our website.