Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Benzalkonium Chloride, 17% (w/v), Solution, Spectrum™ Chemical
SDP

Catalog No. 18600880 Shop All Spectrum Chemical Manufacturing Corporation Products
Click to view available options
Quantity:
500 mL
4 L
200 L
Packaging:
Amber Glass Bottle
Drum

B1069, 8001-54-5

Spectrum™ Chemical Benzalkonium Chloride, 17 Percent (w/v), Solution is also referred to as BZK is an aqueous solution for handling purposes. It varies in color, from colorless to a pale yellow and has an almond-like smell and bitter taste. Its uses vary from disinfectant to microbial corrosion inhibition and can also be used as an algaecide.

Chemical Identifiers

CAS 8001-54-5, 7732-18-5
Molecular Formula C9H13ClN
Molecular Weight (g/mol) 170.66
SMILES [Cl-].C[N+](C)(*)CC1=CC=CC=C1

Specifications

CAS Max % 17%, 83%
Packaging Amber Glass Bottle
Quantity 4 L
UN Number UN1760
Percent Purity 17% ±1.5
Grade Chemical Solution
Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Your feedback has been submitted: Thank you for helping us improve our website.