Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More
Learn More
Benzalkonium Chloride, 95-104%, Spectrum™ Chemical

Click to view available options
Quantity:
100 g
1 kg
25 kg
Packaging:
Amber Glass Bottle
Fiber Drum
Glass Bottle
Description
Spectrum™ Chemical Benzalkonium Chloride is used as a biocide, a phase transfer agent, and a cationic surfactant in the chemical industry. Ungraded products supplied by Spectrum are indicative of a grade suitable for general industrial use or research purposes and typically are not suitable for human consumption or therapeutic use. These materials may or may not have a Certificate of Analysis available.
Specifications
Specifications
CAS Max % | 100% |
Quantity | 1 kg |
UN Number | UN2923 |
Percent Purity | 95 to 104% |
Grade | Reagent |
Packaging | Glass Bottle |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction