Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Benzalkonium chloride

Catalog No. AA4133922
Click to view available options
Quantity:
25 g
100 g
500 g

Cationic surface active agent | CAS: 63449-41-2 | C21H38ClN | 339.99 g/mol

Cationic surface active agentBenzalkonium chloride is used in textile dyeing, softeners, antistatic agents, emulsifiers and conditioner. It acts as a cationic detergent. It is also used as a preservative in pharmaceutical products like eye, ear and nasal drops and sprays. Further, it is an active ingredient in personal care products such as hand sanitizers, shampoos, deodorants and wet wipes. In addition to this, it is used in a study to assess its action on epithelia conjunctival cells in vitro.

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

  • Ampouled under argon
TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 63449-41-2
Molecular Formula C21H38ClN
Molecular Weight (g/mol) 339.99
MDL Number MFCD00137276
InChI Key JBIROUFYLSSYDX-UHFFFAOYSA-M
Synonym benzyldimethyldecylammonium chloride, benzenemethanaminium, n-decyl-n,n-dimethyl-, chloride, benzenemethanaminium, n-decyl-n,n-dimethyl-, chloride 1:1, quaternary ammonium compounds, benzyl-c8-18-alkyldimethyl, chlorides, roccal, benzyl decyl dimethylazanium chloride, tret-o-lite xc 511, ccris 4586, benzyl decyl dimethylammonium chloride, c8-18-alkydimethylbenzyl ammonium chlorides
PubChem CID 13762
IUPAC Name benzyl-decyl-dimethylazanium;chloride
SMILES [Cl-].CCCCCCCCCCCC[N+](C)(C)CC1=CC=CC=C1

Specifications

Quantity 100 g
Solubility Information Miscible with water,acetone,ether and alcohol. Slightly miscible with benzene.
Physical Form Semi-solid
Chemical Name or Material Benzalkonium Chloride
Hazard Category H302+H312-H314-H335
Precautionary Statement P260-P264b-P270-P271-P280-P303+P361+P353-P304+P340-P305+P351+P338-P310-P330-P331-P363-P501c
DOTInformation Hazard Class: 8; Packaging Group: III
EINECSNumber 264-151-6
TSCA Yes
Recommended Storage Ambient temperatures

RUO – Research Use Only

Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Your feedback has been submitted: Thank you for helping us improve our website.