Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Benzalkonium chloride, alkyl distribution C8-C16, 50 wt% aqueous solution

Catalog No. ac263820010
Click to view available options
Quantity:
1 L
2.5 L
Packaging:
Glass bottle

CAS: 8001-54-5 | C9H13ClN | 170.66 g/mol

This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 8001-54-5, 7732-18-5
Molecular Formula C9H13ClN
Molecular Weight (g/mol) 170.66
MDL Number MFCD00145757
Synonym Alkyl-benzyl-dimethylammonium chloride

Specifications

Color Colorless to Yellow
Boiling Point >100.0°C
CAS Min % 49.0, 48.0
CAS Max % 52.0, 51.0
Linear Formula C6H5CH2N(CH3)2RClR=C8H17toC18H37
Concentration 49 to 52% (based on M.W.= 352.5)
Density 0.9800g/mL
Merck Index 15, 1061
Solubility Information Solubility in water: soluble. Other solubilities: soluble in alcohol and acetone, slightly soluble in benzene, almost insoluble in ether
Packaging Glass bottle
Specific Gravity 0.98
Viscosity 130 mPa.s (20°C)
pH 6 to 9 (10% aq. soln.)
Quantity 1 L
Flash Point 105°C
Physical Form Solution
Chemical Name or Material Benzalkonium chloride
Show More Show Less
Hazard Category Danger
Hazard Statement GHS P Statement Causes severe skin burns and eye damage. Harmful if swallowed. Very toxic to aquatic life with long lasting effects.
Precautionary Statement GHS P Statement IF SWALLOWED: rinse mouth. Do NOT induce vomiting. Wear eye protection/face protection. IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing. Immediately call a POISON CENTER or doctor/physician. Avoid release to the environment.

RUO – Research Use Only

Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Your feedback has been submitted: Thank you for helping us improve our website.