Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Benzamidine hydrochloride hydrate, 98%, water ca 10-14%

Catalog No. aaa1600206
Click to view available options
Quantity:
5 g
25 g
100 g

CAS: 206752-36-5 | C7H9ClN2 | 156.61 g/mol

Benzamidine hydrochloride hydrate is a reversible inhibitor of trypsin, trypsin-like enzymes, and serine proteases. It is a proteolytic inhibitor and apoptosis reagent.

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Applications
Benzamidine hydrochloride hydrate is a reversible inhibitor of trypsin, trypsin-like enzymes, and serine proteases. It is a proteolytic inhibitor and apoptosis reagent.

Solubility
Soluble in water.

Notes
Keep container tightly sealed. Store in cool, dry conditions in well sealed containers. Incompatible with oxidizing agents. It is hygroscopic in nature.
TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 206752-36-5
Molecular Formula C7H9ClN2
Molecular Weight (g/mol) 156.61
MDL Number MFCD00066285
InChI Key LZCZIHQBSCVGRD-UHFFFAOYSA-N
Synonym benzamidine hydrochloride hydrate, benzenecarboximidamide hydrochloride hydrate, benzamidine hydrate hydrochloride, benzamidinium chloride, benzamidinehydrochloridehydrate, amidinobenzene hydrochloride, benzamidine hydrochloride monohydrate, acmc-20a6bw, ksc491m3d, benzamidin hydrochloride hydrate
PubChem CID 16219042
SMILES Cl.NC(=N)C1=CC=CC=C1

Specifications

Melting Point 82°C to 85°C
Quantity 5 g
Beilstein 3594959
Sensitivity Hygroscopic
Solubility Information Soluble in water.
IUPAC Name benzenecarboximidamide;hydrate;hydrochloride
Formula Weight 156.62 (Anhydrous)
Percent Purity 98%
Assay water ≈10 to 14%
Chemical Name or Material Benzamidine hydrochloride hydrate
Hazard Category H315-H319-H335
Hazard Statement GHS H Statement
H315-H319-H335
Causes skin irritation.
Causes serious eye irritation.
May cause respiratory irritation.
Precautionary Statement P261-P264b-P271-P280-P302+P352-P304+P340-P305+P351+P338-P312-P332+P313-P362-P501c
EINECSNumber 216-795-4
RTECSNumber CV6260000
TSCA Yes
Recommended Storage Ambient temperatures; Store under Nitrogen

RUO – Research Use Only

Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Your feedback has been submitted: Thank you for helping us improve our website.