Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Benzene Solution, One analyte, 100μg/mL in methanol, Ultra Scientific

Catalog No. usam1001
Encompass
Change view
Click to view available options
Quantity:
1 x 1 mL
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
US-AM-100-1 1 x 1 mL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. US-AM-100-1 Supplier Agilent Technologies Supplier No. AM1001
Only null left
Add to Cart
Add to Cart

Chemical Identifiers

CAS 71-43-2 , 67-56-1
Molecular Formula C6H6
Molecular Weight (g/mol) 78.11
MDL Number MFCD00003009
InChI Key UHOVQNZJYSORNB-UHFFFAOYSA-N
PubChem CID 241
ChEBI CHEBI:16716
IUPAC Name benzene
SMILES C1=CC=CC=C1

Specifications

Physical Form Liquid
Color Colorless
Quantity 1 x 1 mL
Concentration or Composition (by Analyte or Components) 100μg/mL
Chemical Name or Material Benzene Solution, 100μg/mL
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.