Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Benzenesulfinic acid, sodium salt, 97%

Catalog No. AC154055000
Click to view available options
Quantity:
100 g
500 g

CAS: 873-55-2 | C6H5NaO2S | 164.15 g/mol

This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 873-55-2
Molecular Formula C6H5NaO2S
Molecular Weight (g/mol) 164.15
MDL Number MFCD00013135
InChI Key CHLCPTJLUJHDBO-UHFFFAOYSA-M
Synonym sodium benzenesulfinate, benzenesulfinic acid sodium salt, benzenesulfinic acid, sodium salt, sodium phenylsulfinate, sodium benzenesulphinate, sodium benzene sulfinate, sodiumphenylsulfinate, phso2na, benzenesulfinic acid, sodium salt 1:1, sodium phenylsulphinate
PubChem CID 2723840
IUPAC Name sodium;benzenesulfinate
SMILES C1=CC=C(C=C1)S(=O)[O-].[Na+]

Specifications

CAS Min % 96.0
CAS Max % 100.0
Melting Point >300.0°C
Color White
Infrared Spectrum Authentic
Assay Percent Range 97%
Packaging Plastic bottle
Linear Formula C6H5SO2Na
Quantity 500 g
Beilstein 11, 2
Fieser 06,526
Formula Weight 164.15
Percent Purity 97%
Physical Form Crystalline Powder or Crystals
Chemical Name or Material Benzenesulfinic acid, sodium salt, 97%
Show More Show Less
EINECSNumber 212-842-8
TSCA TSCA

RUO – Research Use Only

Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Your feedback has been submitted: Thank you for helping us improve our website.