Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Benzenesulfinic acid sodium salt, 98%

Catalog No. p-7081870
Click to view available options
Quantity:
25 g
100 g
500 g

CAS: 873-55-2 | C6H5NaO2S | 164.154 g/mol

It is used as a polymer adhesive enhancers, plasticizers for plasticized and modified of polyamide, epoxy resin, phenolic resin. It is designed for manufacturing of photography, pharmaceutical, electroplating industry, can be used as photo-reducing agent.

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Applications
It is used as a polymer adhesive enhancers, plasticizers for plasticized and modified of polyamide, epoxy resin, phenolic resin. It is designed for manufacturing of photography, pharmaceutical, electroplating industry, can be used as photo-reducing agent.

Solubility
Soluble in water.

Notes
Hygroscopic. Incompatible with strong oxidants. Store in cool, dry place in a tightly sealed container.
TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 873-55-2
Molecular Formula C6H5NaO2S
Molecular Weight (g/mol) 164.154
MDL Number MFCD00013135
InChI Key CHLCPTJLUJHDBO-UHFFFAOYSA-M
Synonym sodium benzenesulfinate, benzenesulfinic acid sodium salt, benzenesulfinic acid, sodium salt, sodium phenylsulfinate, sodium benzenesulphinate, sodium benzene sulfinate, sodiumphenylsulfinate, phso2na, benzenesulfinic acid, sodium salt 1:1, sodium phenylsulphinate
PubChem CID 2723840
IUPAC Name sodium;benzenesulfinate
SMILES C1=CC=C(C=C1)S(=O)[O-].[Na+]

Specifications

Melting Point >300°C
Quantity 25 g
Beilstein 3598405
Solubility Information Soluble in water.
Formula Weight 164.16
Percent Purity 98%
Chemical Name or Material Benzenesulfinic acid sodium salt
EINECSNumber 212-842-8
RTECSNumber DA9135000
TSCA Yes
Recommended Storage Ambient temperatures

RUO – Research Use Only

Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Your feedback has been submitted: Thank you for helping us improve our website.