Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Benzenesulfohydroxamic Acid 98.0+%, TCI America™
SDP

Catalog No. p-7130402
Click to view available options
Quantity:
5 g
25 g

Chemical Identifiers

CAS 599-71-3
Molecular Formula C6H7NO3S
Molecular Weight (g/mol) 173.186
MDL Number MFCD00025013
InChI Key BRMDATNYMUMZLN-UHFFFAOYSA-N
Synonym piloty's acid, benzenesulfohydroxamic acid, benzenesulfonamide, n-hydroxy, benzenesulphonohydroxamic acid, benzsulfohydroxamic acid, hydroxamic acid, benzsulfo, n-phenylsulfonyl hydroxylamine, hydroxylamine, n-phenylsulfonyl, unii-c64l4umh19, n-hydroxy-benzenesulfonamide
PubChem CID 69033
IUPAC Name N-hydroxybenzenesulfonamide
SMILES C1=CC=C(C=C1)S(=O)(=O)NO

Specifications

Melting Point 126°C
Color White-Yellow
Quantity 5 g
Formula Weight 173.19
Percent Purity ≥98.0% (HPLC,T)
Physical Form Crystalline Powder
Chemical Name or Material Benzenesulfohydroxamic Acid
RTECSNumber MX9350000
TSCA Yes
Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Your feedback has been submitted: Thank you for helping us improve our website.