Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Benzenesulfonic acid, sodium salt, 98%

Catalog No. AC401855000
Change view
Click to view available options
Quantity:
25 g
100 g
500 g
Packaging:
Glass bottle
3 product options available for selection
Product selection table with 3 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity Packaging
AC401855000 500 g Glass bottle
AC401850250 25 g Glass bottle
AC401851000 100 g Glass bottle
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 3 options available.
3 options
Catalog No. AC401855000 Supplier Thermo Scientific Chemicals Supplier No. 401855000
Add to Cart

missing translation for 'validMainframeMsg'

CAS: 515-42-4 | C6H5NaO3S | 180.16 g/mol

This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 515-42-4
Molecular Formula C6H5NaO3S
Molecular Weight (g/mol) 180.16
MDL Number MFCD00065179
InChI Key MZSDGDXXBZSFTG-UHFFFAOYSA-M
Synonym sodium benzenesulfonate, benzenesulfonic acid sodium salt, benzenesulfonic acid, sodium salt, dispersant gn, sodium phenylsulfonate, sodium benzenesulphonate, sodium benzene sulfonate, sodium benzene monosulfate, unii-k5rm14azhx, sodium benzene monosulfonate
PubChem CID 517327
IUPAC Name sodium;benzenesulfonate
SMILES C1=CC=C(C=C1)S(=O)(=O)[O-].[Na+]

Specifications

CAS Min % 97.5
CAS Max % 100.0
Melting Point 450.0°C
Color White
Loss on Drying 2% max. (105°C)
Infrared Spectrum Authentic
Assay Percent Range 97.5% min. (HPLC)
Packaging Glass bottle
Linear Formula SO3NaC6H5
Quantity 500 g
Merck Index 15,1071
Solubility Information (5% in water) Clear colorless to almost colorless
Formula Weight 180.16
Percent Purity 98%
Physical Form Crystalline Powder
Chemical Name or Material Benzenesulfonic acid, sodium salt, 98%
Show More Show Less
Hazard Category HYGROSCOPIC
EINECSNumber 208-198-2
RTECSNumber DB7350000
TSCA TSCA

RUO – Research Use Only

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.