Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Benzethonium chloride, 97%

Catalog No. AAA161420E
Change view
Click to view available options
Quantity:
100 g
2500 g
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
AAA161420E 2500 g
AAA1614222 100 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. AAA161420E Supplier Thermo Scientific Chemicals Supplier No. A161420E
Add to Cart

CAS: 121-54-0 | C27H42ClNO2 | 448.09 g/mol

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Suitable for battery materials development.

Applications
Benzethonium chloride is used as a surface disinfectant and a preservative in food processing and pharmaceutical industries. It is involved in the determination of sodium dodecyl sulfate using sulphan blue as indicator from the mixture containing other ingredients like sodium chloride and sodium sulfate. It is an effective cationic detergent due to its positively charged hydrophilic part of quaternary ammonium ion. It is an active ingredient in medicaments, deodorants and mouthwashes.

Solubility
Soluble in water.

Notes
Hygroscopic. Light sensitive. Incompatible with strong oxidizing agents.
TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 121-54-0
Molecular Formula C27H42ClNO2
MDL Number MFCD00011742
InChI Key UREZNYTWGJKWBI-UHFFFAOYSA-M
Synonym benzethonium chloride, quatrachlor, hyamine, phemeride, phemerol chloride, benzethoniumchloride, phemithyn, disilyn, kylacol, solamin
PubChem CID 8478
ChEBI CHEBI:31264

Specifications

Melting Point 157°C to 165°C
Odor Odorless
Quantity 2500 g
UN Number UN1759
Beilstein 3898548
Sensitivity Hygroscopic
Merck Index 14,1074
Solubility Information Soluble in water.
SMILES CC(C)(C)CC(C)(C)C1=CC=C(C=C1)OCCOCC[N+](C)(C)CC2=CC=CC=C2.[Cl-]
IUPAC Name benzyl-dimethyl-[2-[2-[4-(2,4,4-trimethylpentan-2-yl)phenoxy]ethoxy]ethyl]azanium;chloride
Molecular Weight (g/mol) 448.088
Formula Weight 448.1
Percent Purity 97%
Chemical Name or Material Benzethonium chloride
Show More Show Less
Hazard Category H301-H314-H335
Hazard Statement GHS H Statement
H314-H318-H302
Causes severe skin burns and eye damage.
Causes serious eye damage.
Harmful if swallowed.
Precautionary Statement P260-P264b-P270-P271-P280-P301+P310-P301+P330+P331-P303+P361+P353-P304+P340-P305+P351+P338-P310-P312-P363-P501c
DOTInformation Transport Hazard Class: 8; Packing Group: III; Proper Shipping Name: CORROSIVE SOLIDS, N.O.S.
EINECSNumber 204-479-9
RTECSNumber BO7175000
TSCA Yes
Recommended Storage Ambient temperatures

RUO – Research Use Only

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.