Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

(Benzhydrylthio)acetic Acid 98.0+%, TCI America™
SDP

Catalog No. B417525G Shop All Tokyo Chemical Industry (TCI) Products
Change view
Click to view available options
Quantity:
5 g
25 g
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
B417525G 25 g
B41755G 5 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. B417525G Supplier TCI America Supplier No. B417525G
Add to Cart
Add to Cart

Chemical Identifiers

CAS 63547-22-8
Molecular Formula C15H13O2S
Molecular Weight (g/mol) 257.33
MDL Number MFCD05262040
InChI Key HTHFEDOFDBZPRX-UHFFFAOYSA-M
Synonym (Diphenylmethylthio)acetic Acid
PubChem CID 2077886
IUPAC Name 2-[(diphenylmethyl)sulfanyl]acetate
SMILES [O-]C(=O)CSC(C1=CC=CC=C1)C1=CC=CC=C1

Specifications

Melting Point 130°C
Color White-Yellow
Quantity 25 g
Formula Weight 258.34
Percent Purity ≥98.0% (GC,T)
Physical Form Crystalline Powder
Chemical Name or Material (Benzhydrylthio)acetic Acid
TSCA No
Recommended Storage Freezer
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.