Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Benzil, 99+%

Catalog No. AC105410025
Click to view available options
Quantity:
100 g
500 g
2.5 kg
Packaging:
Plastic bottle

CAS: 134-81-6 | C14H10O2 | 210.23 g/mol

This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 134-81-6
Molecular Formula C14H10O2
Molecular Weight (g/mol) 210.23
MDL Number MFCD00003080
InChI Key WURBFLDFSFBTLW-UHFFFAOYSA-N
Synonym benzil, diphenylethanedione, dibenzoyl, diphenylglyoxal, bibenzoyl, ethanedione, diphenyl, 1,2-diphenylethanedione, diphenylethane-1,2-dione, glyoxal, diphenyl, diphenyldiketon
PubChem CID 8651
ChEBI CHEBI:51507
IUPAC Name 1,2-diphenylethane-1,2-dione
SMILES O=C(C(=O)C1=CC=CC=C1)C1=CC=CC=C1

Specifications

CAS Min % 99.0
CAS Max % 100.0
Melting Point 94.0°C to 97.0°C
Color White
Boiling Point 346.0°C to 348.0°C
Loss on Drying 0.5% max.
Infrared Spectrum Authentic
Assay Percent Range 99% min. (GC)
Packaging Plastic bottle
Linear Formula C6H5COCOC6H5
Quantity 2.5 kg
Merck Index 15,108
Solubility Information Solubility in water: 0.5g/L (20°C). Other solubilities: soluble in alcohol,ether,chloroform,ethyl,acetate,benzene,toluene and nitrobenzene
Formula Weight 210.23
Percent Purity 99+%
Physical Form Crystalline Powder
Chemical Name or Material Benzil, 99+%
Show More Show Less
Hazard Category GHS Signal Word: Warning
Hazard Statement GHS H Statement
Causes serious eye irritation.
Causes skin irritation.
Precautionary Statement GHS P Statement
IF ON SKIN: Wash with plenty of soap and water.
If eye irritation persists: Get medical advice/attention.
IF INHALED: Remove to fresh air and keep at rest in a position comfortable for breathing.
EINECSNumber 205-157-0
RTECSNumber DD1925000
TSCA TSCA

RUO – Research Use Only

Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Your feedback has been submitted: Thank you for helping us improve our website.