Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Benzil monohydrazone, 98%, Thermo Scientific™

Catalog No. AC105430250
Click to view available options
:
25g; Glass bottle

Specifications

CAS 5344-88-7
CAS Min % 97.5
CAS Max % 100.0
Infrared Spectrum Authentic
Assay Percent Range 98%
Packaging Glass Bottle
Molecular Formula C14H12N2O
Linear Formula C6H5C(=NNH2)COC6H5
Quantity 25g
MDL Number MFCD00007625
Synonym benzil monohydrazone, hydrazonodeoxybenzoin, ethanedione, diphenyl-, monohydrazone, 2-hydrazinylidene-1,2-diphenylethan-1-one, 2e-2-hydrazinylidene-1,2-diphenylethanone, 2-hydrazono-1,2-diphenylethanone, 2z-2-hydrazinylidene-1,2-diphenylethanone, 1, hydrazone, cdqpgwnbsosemz-dtqazkpqsa, 2-hydrazono-1,2-diphenyl-ethanone
InChI Key CDQPGWNBSOSEMZ-DTQAZKPQSA-N
SMILES C1=CC=C(C=C1)C(=NN)C(=O)C2=CC=CC=C2
IUPAC Name (2E)-2-hydrazinylidene-1,2-diphenylethanone
Molecular Weight (g/mol) 224.263
PubChem CID 5538956
Formula Weight 224.26
Percent Purity 98%
Chemical Name or Material Benzil monohydrazone, 98%
Show More Show Less
EINECSNumber 226-292-1

RUO – Research Use Only

Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Your feedback has been submitted: Thank you for helping us improve our website.