Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Benzimidazole-5-carboxylic acid, 98%

Catalog No. AAA1598014
Change view
Click to view available options
Quantity:
10 g
25 g
50 g
250 g
4 product options available for selection
Product selection table with 4 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
AAA1598014 25 g
AAA1598009 10 g
AAA1598018 50 g
AAA1598030 250 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 4 options available.
4 options
Catalog No. AAA1598014 Supplier Thermo Scientific Chemicals Supplier No. A1598014
Add to Cart

CAS: 15788-16-6 | C8H6N2O2 | 162.15 g/mol

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 15788-16-6
Molecular Formula C8H6N2O2
Molecular Weight (g/mol) 162.15
MDL Number MFCD00011555
InChI Key COYPLDIXZODDDL-UHFFFAOYSA-N
Synonym 1h-benzimidazole-5-carboxylic acid, 5-benzimidazolecarboxylic acid, benzimidazole-5-carboxylic acid, 1h-benzo d imidazole-5-carboxylic acid, 1h-benzimidazole-6-carboxylic acid, 5-carboxybenzimidazole, 1h-1,3-benzodiazole-5-carboxylic acid, 3h-benzo d imidazole-5-carboxylic acid, 1h-benzimidazole-5-carboxylicacid, 1h-1,3-benzodiazole-6-carboxylic acid
PubChem CID 459456
SMILES OC(=O)C1=CC=C2N=CNC2=C1

Specifications

Melting Point >300°C
Quantity 25 g
Beilstein 5686
IUPAC Name 3H-benzimidazole-5-carboxylic acid
Formula Weight 162.15
Percent Purity 98%
Chemical Name or Material Benzimidazole-5-carboxylic acid
Hazard Category H315-H319-H335
Hazard Statement GHS H Statement
H315-H319-H335
Causes skin irritation.
Causes serious eye irritation.
May cause respiratory irritation.
Precautionary Statement P261-P264b-P271-P280-P302+P352-P304+P340-P305+P351+P338-P312-P332+P313-P362-P501c
TSCA No
Recommended Storage Ambient temperatures

RUO – Research Use Only

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.