Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Benzo-12-crown 4-Ether 98.0+%, TCI America™
SDP

Catalog No. B12491G Shop All Tokyo Chemical Industry (TCI) Products
Change view
Click to view available options
Quantity:
1 g
5 g
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
B12491G 1 g
B12495G 5 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. B12491G Supplier TCI America Supplier No. B12491G
Add to Cart
Add to Cart

Chemical Identifiers

CAS 14174-08-4
Molecular Formula C12H16O4
Molecular Weight (g/mol) 224.26
MDL Number MFCD00059934
InChI Key OAJNZFCPJVBYHB-UHFFFAOYSA-N
Synonym 2,3-Benzo-1,4,7,10-tetraoxadodec-2-ene
PubChem CID 586075
IUPAC Name 2,3,5,6,8,9-hexahydro-1,4,7,10-benzotetraoxacyclododecine
SMILES C1COCCOC2=CC=CC=C2OCCO1

Specifications

Melting Point 46°C
Color White
Quantity 1 g
Formula Weight 224.26
Percent Purity ≥98.0% (GC)
Physical Form Crystalline Powder
Chemical Name or Material Benzo-12-crown 4-Ether
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.