Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Benzo[b]thiophene 1,1-dioxide, 98%

Catalog No. p-7021651
Click to view available options
Quantity:
5 g
25 g

CAS: 825-44-5 | C8H6O2S | 166.19 g/mol

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 825-44-5
Molecular Formula C8H6O2S
Molecular Weight (g/mol) 166.19
MDL Number MFCD00052178
InChI Key FRJNKYGTHPUSJR-UHFFFAOYSA-N
Synonym benzo b thiophene 1,1-dioxide, benzothiophene 1,1-dioxide, thianaphthene 1,1-dioxide, benzo b thiophene, 1,1-dioxide, thianaphthene-1,1-dioxide, benzo b thiophene s,s-dioxide, benzo b thiole-1,1-dione, benzothiophene sulfone, maybridge1_005454
PubChem CID 69997
IUPAC Name 1-benzothiophene 1,1-dioxide
SMILES O=S1(=O)C=CC2=CC=CC=C12

Specifications

Melting Point 137°C to 138°C
Quantity 5 g
Beilstein 116442
Formula Weight 166.2
Percent Purity 98%
Chemical Name or Material Benzo[b]thiophene 1,1-dioxide
EINECSNumber 212-544-8
TSCA No
Recommended Storage Ambient temperatures

RUO – Research Use Only

Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Your feedback has been submitted: Thank you for helping us improve our website.