Learn More
Benzo[b]thiophene-2-boronic acid, 97%
Description
Benzo[b]thiophene-2-boronic acid is used in the synthesis of phosphorescent sensor for quantification of copper(II) ion, UV promoted phenanthridine syntheses, Preparation of CYP11B1 inhibitors for treatment of cortisol dependent diseases, PDE4 inhibitors, Suzuki-Miyaura cross-coupling reactions.
This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
Benzo[b]thiophene-2-boronic acid is used in the synthesis of phosphorescent sensor for quantification of copper(II) ion, UV promoted phenanthridine syntheses, Preparation of CYP11B1 inhibitors for treatment of cortisol dependent diseases, PDE4 inhibitors, Suzuki-Miyaura cross-coupling reactions.
Solubility
Insoluble in water.
Notes
Store in cool, dry conditions in a sealed container. Incompatible with oxidizing agents.
Specifications
Specifications
| Melting Point | ∼255°C |
| Quantity | 1 g |
| Beilstein | 1637924 |
| Solubility Information | Insoluble in water. |
| IUPAC Name | (1-benzothiophen-2-yl)boronic acid |
| Formula Weight | 178.02 |
| Percent Purity | 98% |
| Chemical Name or Material | Benzo[b]thiophene-2-boronic acid |
Safety and Handling
| Hazard Category | H315-H319-H335 |
| Hazard Statement | GHS H Statement H315-H319-H335 Causes skin irritation. Causes serious eye irritation. May cause respiratory irritation. |
| Precautionary Statement | P261-P264b-P271-P280-P302+P352-P304+P340-P305+P351+P338-P312-P332+P313-P362-P501c |
| TSCA | No |
| Recommended Storage | Ambient temperatures |
RUO – Research Use Only
Your input is important to us. Please complete this form to provide feedback related to the content on this product.