Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Benzo[ghi]perylene, Certified Reference Material, MilliporeSigma™ Supelco™

Catalog No. 111017934 Shop All MilliporeSigma Supelco Products
Click to view available options
Quantity:
10 mg
1 g

This certified reference material (CRM) is produced and certified in accordance with ISO/IEC 17025 and ISO 17034. This CRM is traceable to primary material from an NMI, e.g. NIST or NMIJ.

TRUSTED_SUSTAINABILITY

Specifications

CAS 191-24-2
Product Line TraceCERT
Physical Form Neat
Linear Formula C22H12
MDL Number MFCD00004135
Melting Point 277°C to 279°C
Boiling Point >500°C
Synonym 1,12-Benzoperylene
Quantity 1 g
Formula Weight 276.33
Grade TraceCERT™ Analytical Standard
Show More Show Less

TraceCERT is a registered trademark of Sigma-Aldrich Co. LLC

Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Your feedback has been submitted: Thank you for helping us improve our website.