Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Benzocaine, USP, 98-102%, Spectrum™

Click to view available options
Quantity:
1 g
5 g
25 g
125 g
500 g
2.5 kg
12 kg
50 kg
Packaging:
Amber Glass Bottle
Fiber Drum
Poly Pail
94-09-7
C9H11NO2
165.19
BLFLLBZGZJTVJG-UHFFFAOYSA-N
ethyl 4-aminobenzoate
CCOC(=O)C1=CC=C(N)C=C1
94-09-7
C9H11NO2
165.19
BLFLLBZGZJTVJG-UHFFFAOYSA-N
ethyl 4-aminobenzoate
CCOC(=O)C1=CC=C(N)C=C1

BE130, 94-09-7, C9H11NO2

Spectrum™ Benzocaine, USP is a mild local anesthetic. All Spectrum Chemical USP products are manufactured, packaged and stored under current Good Manufacturing Practices (cGMP) per 21CFR part 211 in FDA registered and inspected facilities.
Packaging Amber Glass Bottle
Ignition Residue 0.001
Loss on Drying 0.01
Quantity 125 g
CAS Max % 1
Percent Purity 98 to 102%