Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Benzodiazepine Mouse, Unconjugated, Clone: HM237, Abnova™

Catalog No. 89339264 Shop All Abnova Corporation Products
Change view
Click to view available options
Quantity:
1 mg
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
89-339-264 1 mg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. 89-339-264 Supplier Abnova Corporation Supplier No. MAB21878
Add to Cart
Add to Cart

Mouse Monoclonal Antibody.

Benzodiazepine Monoclonal antibody specifically detects Benzodiazepine in samples. It is validated for ELISA, LFIA.

Specifications

Antigen Benzodiazepine
Applications ELISA, Lateral Flow
Classification Monoclonal
Clone HM237
Conjugate Unconjugated
Host Species Mouse
Quantity 1 mg
Primary or Secondary Primary
Content And Storage Store at 4°C. For long term storage store at -20°C. Aliquot to avoid repeated freezing and thawing.
Form Liquid
Isotype IgG2b
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.