Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Benzofuroxan, 98%

Catalog No. aal1014409
Click to view available options
Quantity:
10 g
50 g

Catalog No. AAL1014409

Supplier: Thermo Scientific Chemicals L1014409

Only null left
Add to Cart

CAS: 480-96-6 | C6H4N2O2 | 136.11 g/mol

Benzofuroxan is used as an oxidant for the preparation of 2-arylsubstituted benzimidazoles, quinoxalines and 1,4-bis(alkylamino)-9,10-anthracenediones. It is also sued in the synthesis of 1,2-dinitrosobenzene.

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Applications
Benzofuroxan is used as an oxidant for the preparation of 2-arylsubstituted benzimidazoles, quinoxalines and 1,4-bis(alkylamino)-9,10-anthracenediones. It is also sued in the synthesis of 1,2-dinitrosobenzene.

Solubility
Soluble in methanol. Insoluble in water.

Notes
Incompatible with strong oxidizing agents.
TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 480-96-6
Molecular Formula C6H4N2O2
Molecular Weight (g/mol) 136.11
MDL Number MFCD00005807
InChI Key OKEAMBAZBICIFP-UHFFFAOYSA-N
Synonym benzofuroxan, benzofuroxane, benzofurazan 1-oxide, benzofurazan oxide, 2,1,3-benzoxadiazole 1-oxide, 2,1,3-benzoxadiazole, 1-oxide, benzofurazan, 1-oxide, 2,1,3-benzoxadiazol-1-ium-1-olate, benzo c 1,2,5-oxadiazol-1-ol, benzofurazan n-oxide
PubChem CID 68075
IUPAC Name 3-oxido-2,1,3-benzoxadiazol-3-ium
SMILES C1=CC2=NO[N+](=C2C=C1)[O-]

Specifications

Melting Point 67°C to 71°C
Quantity 10 g
Beilstein 125112
Solubility Information Soluble in methanol. Insoluble in water.
Formula Weight 136.11
Percent Purity 98%
Chemical Name or Material Benzofuroxan
EINECSNumber 207-559-1
TSCA Yes
Recommended Storage Ambient temperatures

RUO – Research Use Only

Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Your feedback has been submitted: Thank you for helping us improve our website.